N-Boc-ethylendiamin

Lieferant: BACHEM BIOCHEMICA

Synonyme: t-boc-N-EDA

4028304.0001 4028304.0005
BACE4028304.0001EA 0 EUR
BACE4028304.0001 BACE4028304.0005
N-Boc-ethylendiamin
N-Boc-ethylendiamin
Formel: C₇H₁₆N₂O₂
Molecular Weight: 160.22 g/mol
MDL: MFCD00191871
CAS-Nummer: 57260-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR