N-Boc-ethylendiamin

Lieferant: COMBI-BLOCKS

Synonyme: t-boc-N-EDA

AM-1613-25G AM-1613-5G
COBBAM-1613-25GEA 50.7 EUR
COBBAM-1613-25G COBBAM-1613-5G
N-Boc-ethylendiamin
N-Boc-ethylendiamin
Formel: C₇H₁₆N₂O₂
Molecular Weight: 160.22 g/mol
MDL: MFCD00191871
CAS-Nummer: 57260-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR