N-Boc-ethylendiamin
Lieferant: Thermo Scientific
Synonyme:
t-boc-N-EDA
AW00372.250
AW00372DA
MAYBAW00372.250EA
0
EUR
MAYBAW00372.250
MAYBAW00372DA
N-Boc-ethylendiamin
N-Boc-ethylendiamin
Formel:
C₇H₁₆N₂O₂ Molecular Weight: 160.22 g/mol |
MDL:
MFCD00191871 CAS-Nummer: 57260-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...