N-Boc-ethylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
t-boc-N-EDA
15369-1G
15369-25G
15369-5G
SIAL15369-1GEA
132
EUR
SIAL15369-1G
SIAL15369-25G
SIAL15369-5G
N-Boc-ethylendiamin, Sigma-Aldrich®
N-Boc-ethylendiamin
Formel:
C₇H₁₆N₂O₂ Molecular Weight: 160.22 g/mol |
MDL:
MFCD00191871 CAS-Nummer: 57260-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...