Methylglycolat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Hydroxyacetic Acid Methyl Ester, Methyl 2-hydroxyacetate, Glycolic Acid Methyl Ester, Methyl Hydroxyacetate

A17870.06 A17870.14 A17870.22
A17870.06EA 52 EUR
A17870.06 A17870.14 A17870.22
Methylglycolat ≥98%
Methylglycolat

Purity: 98% CAS No.: 96-35-5 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 67°(152°F) Refractive Index: 1.4170 Boiling Point: 150-151° Beilstein Reference: 1699571 MDL No.: MFCD00004667

Formel: C₃H₆O₃
Molecular Weight: 90,08 g/mol
Siedepunkt: 150…151 °C
Dichte: 1,184
Flammpunkt: 67 °C (152 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004667
CAS-Nummer: 96-35-5
EINECS: 202-502-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR