Ethylphenoxyacetat ≥99%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 2-phenoxyacetate
A14156.09
A14156.18
A14156.30
A14156.09EA
32.5
EUR
A14156.09
A14156.18
A14156.30
Ethylphenoxyacetat ≥99%
Ethylphenoxyacetat
Purity: 99%
CAS No.: 2555-49-9
Molecular Formula: C6H5OCH2CO2CH2CH3
Formula Weight: 180.20
Flash Point:
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,20 g/mol Siedepunkt: 251…253 °C Dichte: 1,108 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026895 CAS-Nummer: 2555-49-9 EINECS: 219-867-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...