Ethylphenoxyacetat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-phenoxyacetate

001014-25G 001014-10G
FLUO001014-25GEA 63.9 EUR
FLUO001014-25G FLUO001014-10G
Ethylphenoxyacetat
Ethylphenoxyacetat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,20 g/mol
MDL: MFCD00026895
CAS-Nummer: 2555-49-9
EINECS: 219-867-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR