2'-(Trifluormethoxy)acetanilid

Lieferant: FLUOROCHEM

007529-5G 007529-25G
FLUO007529-5GEA 69.8 EUR
FLUO007529-5G FLUO007529-25G
2'-(Trifluormethoxy)acetanilid
2'-(Trifluormethoxy)acetanilid
Formel: C₉H₈F₃NO₂
MDL: MFCD00204176
CAS-Nummer: 3832-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR