4'-(Trifluormethoxy)acetanilid
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethoxy)acetanilide
007530-5G
007530-25G
FLUO007530-5GEA
45.8
EUR
FLUO007530-5G
FLUO007530-25G
4'-(Trifluormethoxy)acetanilid
4'-(Trifluormethoxy)acetanilid
Formel:
C₉H₈F₃NO₂ Molecular Weight: 219.16 g/mol |
MDL:
MFCD00075293 CAS-Nummer: 1737-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...