3-(N-Boc-amino)benzotrifluorid
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Trifluoromethylphenyl)-carbamic acid tert-butyl ester
AN-2542-5G
AN-2542-1G
COBBAN-2542-5GEA
563
EUR
COBBAN-2542-5G
COBBAN-2542-1G
3-(N-Boc-amino)benzotrifluorid
3-(N-Boc-amino)benzotrifluorid
Formel:
C₁₂H₁₄F₃NO₂ |
MDL:
MFCD11040633 CAS-Nummer: 109134-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...