2'-Fluor-4'-iodacetanilid
Lieferant: COMBI-BLOCKS
AN-1249-5G
AN-1249-1G
COBBAN-1249-5GEA
0
EUR
COBBAN-1249-5G
COBBAN-1249-1G
2'-Fluor-4'-iodacetanilid
2'-Fluor-4'-iodacetanilid
Formel:
C₈H₇FINO |
MDL:
MFCD04039298 CAS-Nummer: 97760-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...