Pentafluorphenyl-1-methansulfonat

Lieferant: BIONET RESEARCH

1Y-0285.1 1Y-0285.10 1Y-0285.5 1Y-0285.500
BIOT1Y-0285.1EA 0 EUR
BIOT1Y-0285.1 BIOT1Y-0285.10 BIOT1Y-0285.5 BIOT1Y-0285.500
Pentafluorphenyl-1-methansulfonat
Pentafluorphenyl-1-methansulfonat
Formel: C₇H₃O₃F₅S
MDL: MFCD06659544
CAS-Nummer: 161912-36-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR