9-Ethylcarbazol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 9-Ethyl-9H-carbazole, N-Ethylcarbazole

A11653.0B A11653.22 A11653.30
A11653.0BEA 262 EUR
A11653.0B A11653.22 A11653.30
9-Ethylcarbazol ≥99%
9-Ethylcarbazole

Purity: 99% CAS No.: 86-28-2 Molecular Formula: C14H13N Formula Weight: 195.27 Flash Point: 186°(367°F) Melting Point: 67-71° Boiling Point: 166°/4mm Beilstein Reference: 148115 MDL No.: MFCD00004967

Formel: C₁₄H₁₃N
Molecular Weight: 195,27 g/mol
Siedepunkt: 166 °C (4 mmHg)
Schmelzpunkt: 67…71 °C
Flammpunkt: 186 °C (367 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004967
CAS-Nummer: 86-28-2
EINECS: 201-660-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR