3-Amino-3-(2-bromphenyl)propionsäure

Lieferant: BIONET RESEARCH

7R-0619.5 7R-0619.500 7R-0619.1
BIOT7R-0619.5EA 0 EUR
BIOT7R-0619.5 BIOT7R-0619.500 BIOT7R-0619.1
3-Amino-3-(2-bromphenyl)propionsäure
3-Amino-3-(2-bromphenyl)propionsäure
Formel: C₉H₁₀BrNO₂
MDL: MFCD01871297
CAS-Nummer: 117391-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR