3-Amino-3-(4-bromphenyl)propionsäure
Lieferant: BIONET RESEARCH
6R-0633.500
6R-0633.1
6R-0633.5
BIOT6R-0633.500EA
0
EUR
BIOT6R-0633.500
BIOT6R-0633.1
BIOT6R-0633.5
3-Amino-3-(4-bromphenyl)propionsäure
3-Amino-3-(4-bromphenyl)propionsäure
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244,0900 g/mol |
MDL:
MFCD00187112 CAS-Nummer: 39773-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...