3-Amino-3-(2,3-dichlorphenyl)propionsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-3-(2,3-dichlorophenyl)propanoic acid

6Y-0854.1 6Y-0854.10 6Y-0854.5 6Y-0854.500
BIOT6Y-0854.1EA 0 EUR
BIOT6Y-0854.1 BIOT6Y-0854.10 BIOT6Y-0854.5 BIOT6Y-0854.500
3-Amino-3-(2,3-dichlorphenyl)propionsäure
3-Amino-3-(2,3-dichlorphenyl)propionsäure
Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234.0800 g/mol
MDL: MFCD01871301
CAS-Nummer: 117391-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR