3-Amino-3-(3,4-dichlorphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(3,4-dichlorophenyl)propanoic acid
7R-0620.500
7R-0620.1
7R-0620.5
BIOT7R-0620.500EA
0
EUR
BIOT7R-0620.500
BIOT7R-0620.1
BIOT7R-0620.5
3-Amino-3-(3,4-dichlorphenyl)propionsäure
3-Amino-3-(3,4-dichlorphenyl)propionsäure
Formel:
C₉H₉Cl₂NO₂ |
MDL:
MFCD00462884 CAS-Nummer: 117391-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...