3-Amino-3-(3-chlorphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(3-chlorophenyl)propanoic acid
6R-0641.500
6R-0641.1
6R-0641.5
BIOT6R-0641.500EA
0
EUR
BIOT6R-0641.500
BIOT6R-0641.1
BIOT6R-0641.5
3-Amino-3-(3-chlorphenyl)propionsäure
3-Amino-3-(3-chlorphenyl)propionsäure
Formel:
C₉H₁₀ClNO₂ Molecular Weight: 199.64 g/mol |
MDL:
MFCD00463160 CAS-Nummer: 68208-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...