3-Amino-3-(4-chlorphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid
3T-0623.500
3T-0623.1
3T-0623.5
BIOT3T-0623.500EA
0
EUR
BIOT3T-0623.500
BIOT3T-0623.1
BIOT3T-0623.5
3-Amino-3-(4-chlorphenyl)propionsäure
3-Amino-3-(4-chlorphenyl)propionsäure
Formel:
C₉H₁₀ClNO₂ Molecular Weight: 199.64 g/mol |
MDL:
MFCD00126370 CAS-Nummer: 19947-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...