3-Amino-3-(4-chlorphenyl)propionsäure
Lieferant: Thermo Scientific
Synonyme: 3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid
KM08205.1 KM08205.10
MAYBKM08205.1EA 0 EUR
MAYBKM08205.1 MAYBKM08205.10
3-Amino-3-(4-chlorphenyl)propionsäure
3-Amino-3-(4-chlorphenyl)propionsäure
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199.64 g/mol
MDL: MFCD00126370
CAS-Nummer: 19947-39-8