3-Amino-3-(2-nitrophenyl)propionsäure 98%

Lieferant: Thermo Scientific

Synonyme: 3-Amino-3-(2-nitrophenyl)propanoic acid

B22176.06 B22176.14 B22176.22
B22176.06EA 58 EUR
B22176.06 B22176.14 B22176.22
3-Amino-3-(2-nitrophenyl)propionsäure 98%
3-Amino-3-(2-nitrophenyl)propionsäure

Purity: 98% CAS No.: 5678-48-8 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 MDL No.: MFCD00090356

Formel: C₉H₁₀N₂O₄
Molecular Weight: 210,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090356
CAS-Nummer: 5678-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR