3-Amino-3-(2-nitrophenyl)propionsäure

Lieferant: Thermo Scientific

Synonyme: 3-Amino-3-(2-nitrophenyl)propanoic acid

KM08101.1 KM08101.10
MAYBKM08101.1EA 0 EUR
MAYBKM08101.1 MAYBKM08101.10
3-Amino-3-(2-nitrophenyl)propionsäure
3-Amino-3-(2-nitrophenyl)propionsäure
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210.19 g/mol
MDL: MFCD00090356
CAS-Nummer: 5678-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR