3-Amino-3-(3-nitrophenyl)propionsäure
Lieferant: BIONET RESEARCH
8R-0617.500
8R-0617.1
8R-0617.5
BIOT8R-0617.500EA
0
EUR
BIOT8R-0617.500
BIOT8R-0617.1
BIOT8R-0617.5
3-Amino-3-(3-nitrophenyl)propionsäure
3-Amino-3-(3-nitrophenyl)propionsäure
Formel:
C₉H₁₀N₂O₄ |
MDL:
MFCD00472612 CAS-Nummer: 5678-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...