3-Amino-3-(4-ethoxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

6R-0617.500 6R-0617.1 6R-0617.5
BIOT6R-0617.500EA 0 EUR
BIOT6R-0617.500 BIOT6R-0617.1 BIOT6R-0617.5
3-Amino-3-(4-ethoxyphenyl)propionsäure
3-Amino-3-(4-ethoxyphenyl)propionsäure
Formel: C₁₁H₁₅NO₃
MDL: MFCD00448268
CAS-Nummer: 38499-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR