3-Amino-3-(4-methoxyphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(4-methoxyphenyl)propionic Acid, 3-Amino-3-(4-methoxyphenyl)propanoic acid
7R-0624.500
7R-0624.1
BIOT7R-0624.500EA
0
EUR
BIOT7R-0624.500
BIOT7R-0624.1
3-Amino-3-(4-methoxyphenyl)propionsäure
3-Amino-3-(4-methoxyphenyl)propionsäure
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,2151 g/mol |
MDL:
MFCD00187209 CAS-Nummer: 5678-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...