3-Amino-3-(3,4-dimethoxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

7R-0605.500 7R-0605.1 7R-0605.5
BIOT7R-0605.500EA 0 EUR
BIOT7R-0605.500 BIOT7R-0605.1 BIOT7R-0605.5
3-Amino-3-(3,4-dimethoxyphenyl)propionsäure
3-Amino-3-(3,4-dimethoxyphenyl)propionsäure
Formel: C₁₁H₁₅NO₄
MDL: MFCD00735172
CAS-Nummer: 696641-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR