3-Amino-3-(3-hydroxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

8R-0664.500 8R-0664.1 8R-0664.5
BIOT8R-0664.500EA 0 EUR
BIOT8R-0664.500 BIOT8R-0664.1 BIOT8R-0664.5
3-Amino-3-(3-hydroxyphenyl)propionsäure
3-Amino-3-(3-hydroxyphenyl)propionsäure
Formel: C₉H₁₁NO₃
MDL: MFCD01871309
CAS-Nummer: 26049-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR