3-Amino-3-(4-hydroxyphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(4-hydroxyphenyl)propanoic Acid
8R-0644.500
8R-0644.1
8R-0644.5
BIOT8R-0644.500EA
0
EUR
BIOT8R-0644.500
BIOT8R-0644.1
BIOT8R-0644.5
3-Amino-3-(4-hydroxyphenyl)propionsäure
3-Amino-3-(4-hydroxyphenyl)propionsäure
Formel:
C₉H₁₁NO₃ Molecular Weight: 181.1885 g/mol |
MDL:
MFCD00181810 CAS-Nummer: 6049-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...