3-(3-Fluor-4-methoxybenzoyl)propionsäure

Lieferant: FLUOROCHEM

009213-5G 009213-25G 009213-1G
FLUO009213-5GEA 0 EUR
FLUO009213-5G FLUO009213-25G FLUO009213-1G
3-(3-Fluor-4-methoxybenzoyl)propionsäure
3-(3-Fluor-4-methoxybenzoyl)propionsäure
Formel: C₁₁H₁₁FO₄
Molecular Weight: 226.20 g/mol
MDL: MFCD00020538
CAS-Nummer: 347-63-7
EINECS: 206-474-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR