3-Amino-3-(3-fluorphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-Amino-3-(3-fluorophenyl)propanoic acid
7R-0608.500
7R-0608.1
7R-0608.5
BIOT7R-0608.500EA
0
EUR
BIOT7R-0608.500
BIOT7R-0608.1
BIOT7R-0608.5
3-Amino-3-(3-fluorphenyl)propionsäure
3-Amino-3-(3-fluorphenyl)propionsäure
Formel:
C₉H₁₀FNO₂ Molecular Weight: 183,1796 g/mol |
MDL:
MFCD00764457 CAS-Nummer: 117391-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...