3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
Lieferant: CHEMPUR SILANE
SILS21126.10
SILS21126.25
SILS21126.100
SILNSILS21126.10EA
0
EUR
SILNSILS21126.10
SILNSILS21126.25
SILNSILS21126.100
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
Formel:
C₁₆H₃₈O₅Si₄ |
MDL:
MFCD00053871 CAS-Nummer: 17096-07-0 EINECS: 241-165-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...