3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat

Lieferant: CHEMPUR SILANE

Synonyme: 3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl methacrylate, 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane

SILS21127.10 SILS21127.25 SILS21127.100
SILNSILS21127.10EA 0 EUR
SILNSILS21127.10 SILNSILS21127.25 SILNSILS21127.100
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
Formel: C₁₆H₃₈O₅Si₄
Molecular Weight: 422,82 g/mol
MDL: MFCD00053871
CAS-Nummer: 17096-07-0
EINECS: 241-165-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR