4-Acetoxy-3-methoxyphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
010807-5G
010807-1G
FLUO010807-5GEA
602
EUR
FLUO010807-5G
FLUO010807-1G
4-Acetoxy-3-methoxyphenylboronsäurepinakolester
4-Acetoxy-3-methoxyphenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₅ |
MDL:
MFCD03789257 CAS-Nummer: 811841-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...