4-Methoxy-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Methoxy-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5083-1G PN-5083-250M
COBBPN-5083-1GEA 742 EUR
COBBPN-5083-1G COBBPN-5083-250M
4-Methoxy-2-nitrophenylboronsäurepinakolester
4-Methoxy-2-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₅
Molecular Weight: 279,1000 g/mol
MDL: MFCD10699701
CAS-Nummer: 1073353-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR