4-Methoxy-3-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-Methoxy-3-nitrophenylboronic acid, pinacol ester, 2-(4-Methoxy-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-5076-5G
PN-5076-1G
COBBPN-5076-5GEA
884
EUR
COBBPN-5076-5G
COBBPN-5076-1G
4-Methoxy-3-nitrophenylboronsäurepinakolester
4-Methoxy-3-nitrophenylboronsäurepinakolester
Formel:
C₁₃H₁₈BNO₅ Molecular Weight: 279.0967 g/mol |
MDL:
MFCD06795663 CAS-Nummer: 554411-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...