4-Hydroxy-3-methoxyphenylboronsäurepinakolester 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
367540010
367540050
ACRO367540010EA
72.5
EUR
ACRO367540010
ACRO367540050
4-Hydroxy-3-methoxyphenylboronsäurepinakolester 97%
4-Hydroxy-3-methoxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₉BO₄ Molecular Weight: 250,099 g/mol Schmelzpunkt: 101…106 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093723 CAS-Nummer: 269410-22-2 |
Spezifikation Testergebnisse
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 101 to 106 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...