4-Hydroxy-3-methoxyphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

010806-5G FL10806.20 010806-1G
FLUO010806-5GEA 525 EUR
FLUO010806-5G CHMPFL10806.20 FLUO010806-1G
4-Hydroxy-3-methoxyphenylboronsäurepinakolester
4-Hydroxy-3-methoxyphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₄
Molecular Weight: 250.099 g/mol
MDL: MFCD02093723
CAS-Nummer: 269410-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR