2-Formyl-4-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2300-5G
BB-2300-1G
COBBBB-2300-5GEA
352
EUR
COBBBB-2300-5G
COBBBB-2300-1G
2-Formyl-4-methoxyphenylboronsäure
2-Formyl-4-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179.9657 g/mol |
MDL:
MFCD01319043 CAS-Nummer: 139962-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...