2-Formyl-4-methoxyphenylboronsäure

Lieferant: FLUOROCHEM

011046-5G 011046-1G
FLUO011046-5GEA 350 EUR
FLUO011046-5G FLUO011046-1G
2-Formyl-4-methoxyphenylboronsäure
2-Formyl-4-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.9657 g/mol
MDL: MFCD01319043
CAS-Nummer: 139962-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR