2-Formyl-5-methoxyphenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
5-Methoxy-2-formylphenylboronic acid
011047-5G
011047-1G
FLUO011047-5GEA
495
EUR
FLUO011047-5G
FLUO011047-1G
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179,9657 g/mol |
MDL:
MFCD03001335 CAS-Nummer: 40138-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...