3,5-Diformyl-2-isopropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8085-1G BB-8085-250M
COBBBB-8085-1GEA 651 EUR
COBBBB-8085-1G COBBBB-8085-250M
3,5-Diformyl-2-isopropoxyphenylboronsäure
3,5-Diformyl-2-isopropoxyphenylboronsäure
Formel: C₁₁H₁₃BO₅
MDL: MFCD09750451
CAS-Nummer: 1072951-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR