4-Methoxy-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methoxy-2-nitrophenyl)boronic acid

BB-5083-1G BB-5083-250M
COBBBB-5083-1GEA 1400 EUR
COBBBB-5083-1G COBBBB-5083-250M
4-Methoxy-2-nitrophenylboronsäure
4-Methoxy-2-nitrophenylboronsäure
Formel: C₇H₈BNO₅
Molecular Weight: 196,950 g/mol
MDL: MFCD09878340
CAS-Nummer: 860034-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR