3-(Trimethylsilyl)phenylboronsäure
Lieferant: CHEMPUR SILANE
Synonyme:
(3-(Trimethylsilyl)phenyl)boronic acid
SILS20000.1
SILS20000.5
SILNSILS20000.1EA
0
EUR
SILNSILS20000.1
SILNSILS20000.5
3-(Trimethylsilyl)phenylboronsäure
3-(Trimethylsilyl)phenylboronsäure
Formel:
C₉H₁₅BO₂Si Molecular Weight: 194,1100 g/mol |
MDL:
MFCD03093884 CAS-Nummer: 177171-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...