4-(Trimethylsilyl)phenylboronsäure
Lieferant: CHEMPUR SILANE
Synonyme:
(4-(Trimethylsilyl)phenyl)boronic acid
SILS20001.1
SILS20001.5
SILNSILS20001.1EA
0
EUR
SILNSILS20001.1
SILNSILS20001.5
4-(Trimethylsilyl)phenylboronsäure
4-(Trimethylsilyl)phenylboronsäure
Formel:
C₉H₁₅BO₂Si Molecular Weight: 194.1100 g/mol |
MDL:
MFCD00093348 CAS-Nummer: 17865-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...