Kalium(2-formylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium trifluoro(2-formylphenyl)borate, Potassium 2-formylphenyltrifluoroborate

TB-3237-1G TB-3237-25G TB-3237-5G
COBBTB-3237-1GEA 37.5 EUR
COBBTB-3237-1G COBBTB-3237-25G COBBTB-3237-5G
Kalium(2-formylphenyl)trifluoroborat
Kalium(2-formylphenyl)trifluoroborat
Formel: C₇H₅BF₃KO
Molecular Weight: 212,02 g/mol
MDL: MFCD02093979
CAS-Nummer: 192863-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR