Kalium(2-(aminocarbonyl)phenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
Potassium (2-carbamoylphenyl)trifluoroborate
TB-3240-1G
TB-3240-5G
COBBTB-3240-1GEA
393
EUR
COBBTB-3240-1G
COBBTB-3240-5G
Kalium(2-(aminocarbonyl)phenyl)trifluoroborat
Kalium(2-(aminocarbonyl)phenyl)trifluoroborat
Formel:
C₇H₆BF₃KNO |
MDL:
MFCD04115765 CAS-Nummer: 850623-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...