Kalium(4-(aminocarbonyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium 4-carbamoylphenyltrifluoroborate, Potassium (4-carbamoylphenyl)trifluoroborate

TB-3230-5G TB-3230-25G TB-3230-1G
COBBTB-3230-5GEA 302 EUR
COBBTB-3230-5G COBBTB-3230-25G COBBTB-3230-1G
Kalium(4-(aminocarbonyl)phenyl)trifluoroborat
Kalium(4-(aminocarbonyl)phenyl)trifluoroborat
Formel: C₇H₆BF₃KNO
Molecular Weight: 227,03 g/mol
MDL: MFCD04115758
CAS-Nummer: 850623-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR