Kalium(2-(ethoxycarbonyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium (2-(ethoxycarbonyl)phenyl)trifluoroborate

TB-3244-1G TB-3244-5G
COBBTB-3244-1GEA 204 EUR
COBBTB-3244-1G COBBTB-3244-5G
Kalium(2-(ethoxycarbonyl)phenyl)trifluoroborat
Kalium(2-(ethoxycarbonyl)phenyl)trifluoroborat
Formel: C₉H₉BF₃KO₂
Molecular Weight: 256,0711 g/mol
MDL: MFCD04115768
CAS-Nummer: 850623-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR