Kalium(3-carboxy-5-nitrophenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3249-1G TB-3249-5G
COBBTB-3249-1GEA 49.7 EUR
COBBTB-3249-1G COBBTB-3249-5G
Kalium(3-carboxy-5-nitrophenyl)trifluoroborat
Potassium-3-carboxy-5-nitrophenyltrifluoroborate
Formel: C₇H₄BF₃KNO₄
MDL: MFCD04115772
CAS-Nummer: 850623-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR