Kalium(2-(methylthio)phenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
Potassium trifluoro(2-(methylthio)phenyl)borate, Potassium(2-Methylsulphonylphenyl)trifluoroboroate
TB-3250-1G
TB-3250-25G
TB-3250-5G
COBBTB-3250-1GEA
74.2
EUR
COBBTB-3250-1G
COBBTB-3250-25G
COBBTB-3250-5G
Kalium(2-(methylthio)phenyl)trifluoroborat
Potassium-2-(methylthio)phenyltrifluoroborate
Formel:
C₇H₇BF₃KS Molecular Weight: 230,10 g/mol |
MDL:
MFCD04115773 CAS-Nummer: 850623-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...